Anti-MARK1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB2698
10679-250EA 329 USD
10679-250
Anti-MARK1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of MARK1.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:200-500; Immunohistochemistry: 1:10-50

Type: Primary
Antigen: MARK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR