Anti-SIRT6 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12037
10737-204EA 530.32 USD
10737-204
Anti-SIRT6 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of SIRT6.

  • Recommended Dilutions: Western Blot: 0.5-2 μg/ml

Type: Primary
Antigen: SIRT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR