Anti-MSLN Mouse Monoclonal Antibody [clone: MN-1]

Supplier: Abnova
MAB1993
10726-016EA 746.62 USD
10726-016
Anti-MSLN Mouse Monoclonal Antibody [clone: MN-1]
Antibodies
Mouse monoclonal antibody raised against partial recombinant MSLN.

Recommended Dilutions: Western Blot: 1:1000; Immunohistochemistry: 1:100

Type: Primary
Antigen: MSLN
Clonality: Monoclonal
Clone: MN-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR