Anti-S100 A12 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12047
10734-494EA 530.32 USD
10734-494
Anti-S100 A12 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against full length recombinant S100A12.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: S100 A12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR