Anti-ROBO4 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12048
10731-824EA 530.32 USD
10731-824
Anti-ROBO4 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of ROBO4.

Recommended Dilutions: Western Blot: 1:500; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 5 ?g/ml

Type: Primary
Antigen: ROBO4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR