Anti-MTBP Mouse Monoclonal Antibody [clone: PHL-1]

Supplier: Abnova
MAB1995
10726-086EA 746.62 USD
10726-086
Anti-MTBP Mouse Monoclonal Antibody [clone: PHL-1]
Antibodies
Mouse monoclonal antibody raised against partial recombinant MTBP.

Recommended Dilutions: ELISA: 1:2000-1:20000; Western Blot: 1:200-1:2000

Type: Primary
Antigen: MTBP
Clonality: Monoclonal
Clone: PHL-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chimpanzee, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR