Anti-p13 Myeloid Marker Mouse Monoclonal Antibody [clone: BM3]

Supplier: Abnova
MAB1997
10733-306EA 746.62 USD
10733-306
Anti-p13 Myeloid Marker Mouse Monoclonal Antibody [clone: BM3]
Antibodies
Mouse monoclonal antibody raised against p13 Myeloid Marker.

Recommended Dilutions: ELISA: 1:5000-1:20000; Western Blot: 1:500-1:5,000; Immunoprecipitation: 1:100

Type: Primary
Antigen: p13 Myeloid Marker
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: BM3
Epitope:
Host: IgG1 kappa
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR