Anti-RAD50 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12052
10734-094EA 530.32 USD
10734-094
Anti-RAD50 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against partial recombinant RAD50.

Recommended Dilutions: Western Blot: 1:1000-1:10000; Immunoprecipitation: 1-5 ?g/ml lysate)

Type: Primary
Antigen: RAD50
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR