Anti-PEAR1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB22861
10728-836EA 428.55 USD
10728-836
Anti-PEAR1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant PEAR1.

Recommended Dilutions: Immunohistochemistry: 1:20-1:50

Type: Primary
Antigen: PEAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR