Anti-Thyroxine Mouse Monoclonal Antibody [clone: 1H1]

Supplier: Abnova
MAB2885
10677-346EA 329 USD
10677-346
Anti-Thyroxine Mouse Monoclonal Antibody [clone: 1H1]
Antibodies
Mouse monoclonal antibody raised against Thyroxine.

Type: Primary
Antigen: Thyroxine
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 1H1
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR