Anti-D-Dimer Mouse Monoclonal Antibody [clone: DD1]

Supplier: Abnova
MAB2918
10676-128EA 345.2 USD
10676-128
Anti-D-Dimer Mouse Monoclonal Antibody [clone: DD1]
Antibodies
Mouse monoclonal antibody raised against D-Dimer.

Type: Primary
Antigen: D-Dimer
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: DD1
Epitope:
Host: IgG2a
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR