Anti-ACAN Mouse Monoclonal Antibody [clone: 14A6 3B2]

Supplier: Abnova
MAB3670
10626-452EA 1061 USD
10626-452
Anti-ACAN Mouse Monoclonal Antibody [clone: 14A6 3B2]
Antibodies
Mouse monoclonal antibody raised against recombinant ACAN.

Type: Primary
Antigen: ACAN
Clonality: Monoclonal
Clone: 14A6 3B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR