Anti-POU1F1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23836
10729-706EA 428.55 USD
10729-706
Anti-POU1F1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant POU1F1.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000; Immunofluorescence: 1-4 ?g/ml

Type: Primary
Antigen: POU1F1
Clonality: Unconjugated
Clone: Polyclonal
Conjugation:
Epitope:
Host: IgG
Isotype: Rabbit
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR