Anti-LEP Mouse Monoclonal Antibody [clone: 1E9 1H6]

Supplier: Abnova
MAB3685
10628-240EA 1060.83 USD
10628-240
Anti-LEP Mouse Monoclonal Antibody [clone: 1E9 1H6]
Antibodies
Mouse monoclonal antibody raised against recombinant LEP.

Type: Primary
Antigen: LEP
Clonality: Monoclonal
Clone: 1E9 1H6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR