Anti-CSH1 Mouse Monoclonal Antibody [clone: 6/6A3]

Supplier: Abnova
MAB3692
10627-124EA 530.63 USD
10627-124
Anti-CSH1 Mouse Monoclonal Antibody [clone: 6/6A3]
Antibodies
Mouse monoclonal antibody raised against recombinant CSH1.

Type: Primary
Antigen: CSH1
Clonality: Monoclonal
Clone: 6/6A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR