Anti-REN Mouse Monoclonal Antibody [clone: 244/1 RH12]

Supplier: Abnova
MAB3694
10628-646EA 1276.64 USD
10628-646
Anti-REN Mouse Monoclonal Antibody [clone: 244/1 RH12]
Antibodies
Mouse monoclonal antibody raised against recombinant REN.

Type: Primary
Antigen: REN
Clonality: Monoclonal
Clone: 244/1 RH12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR