Anti-DFFA Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13069
10710-580EA 428 USD
10710-580
Anti-DFFA Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of DFFA.

Recommended Dilutions: Western Blot: 1:1000-1:2000; Immunohistochemistry: 1:1000-1:2000

Type: Primary
Antigen: DFFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR