Anti-WNT1 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB11291
10744-426EA 786.15 USD
10744-426
Anti-WNT1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of WNT1.

Recommended Dilutions: ELISA: 1:25000; Western Blot: 1:1500-1:6000

Type: Primary
Antigen: WNT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Chicken, Dog, Human, Macaque, Mouse, Opossum, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR