Anti-RFX5 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB11300
10731-544EA 638 USD
10731-544
Anti-RFX5 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of RFX5.

Recommended Dilutions: Western Blot: 1:1000; Gel Supershift Assay (0.5-1.0 ?l per assay)

Type: Primary
Antigen: RFX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR