Anti-CD46 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-258]

Supplier: Abnova
MAB4439
10706-636EA 329 USD
10706-636
Anti-CD46 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-258]
Antibodies
Mouse monoclonal antibody raised against native CD46.

Recommended Dilutions: Flow Cytometry: 10 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)

Type: Primary
Antigen: CD46
Clonality: Monoclonal
Clone: MEM-258
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR