Anti-RFA2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB11311
10675-292EA 638 USD
10675-292
Anti-RFA2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of RFA2.

Recommended Dilutions: ELISA: 1:8000-1:20000 of phosphorylated form 1:300 of non-phosphorylated form; Western Blot: 1:500-1:1000

Type: Primary
Antigen: RFA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR