Anti-CD52 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HI186]

Supplier: Abnova
MAB4449
10706-676EA 329 USD
10706-676
Anti-CD52 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: HI186]
Antibodies
Mouse monoclonal antibody raised against native CD52.

Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)

Type: Primary
Antigen: CD52
Clonality: Monoclonal
Clone: HI186
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR