Anti-POGZ Rabbit Polyclonal Antibody

Supplier: Abnova
PAB11320
10729-626EA 638 USD
10729-626
Anti-POGZ Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Pogz.

Recommended Dilutions: ELISA: 1:100000-1:400000; Western Blot: 1:2000-1:20000

Type: Primary
Antigen: POGZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Baboon, Bovine, Chicken, Chimpanzee, Dog, Human, Macaque, Mouse, Opossum, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR