Anti-SNAP25 Mouse Monoclonal Antibody [clone: S-7B8]

Supplier: Abnova
MAB2019
10738-350EA 428 USD
10738-350
Anti-SNAP25 Mouse Monoclonal Antibody [clone: S-7B8]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of Snap25.

Recommended Dilutions: Western Blot: 1 to 5 ?g/ml; Immunohistochemistry on paraformaldehyde fixed CNS tissues (1 to 5 ?g/ml

Type: Primary
Antigen: SNAP25
Clonality: Monoclonal
Clone: S-7B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Chicken, Human, Monkey, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR