Anti-STAT5A Rabbit Polyclonal Antibody

Supplier: Abnova
PAB11328
10739-708EA 638 USD
10739-708
Anti-STAT5A Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Stat5a.

Recommended Dilutions: ELISA: 1:25000; Western Blot: 1:500-1:2000

Type: Primary
Antigen: STAT5A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR