Anti-RBL2 Monoclonal Antibody [clone: KAB40]

Supplier: Abnova
MAB2026
10734-162EA 428 USD
10734-162
Anti-RBL2 Monoclonal Antibody [clone: KAB40]
Antibodies
Mouse monoclonal antibody raised against partial recombinant RBL2.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: RBL2
Clonality: Monoclonal
Clone: KAB40
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity: Human, Monkey, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR