Anti-CDKN2D Mouse Monoclonal Antibody [clone: DCS-100]

Supplier: Abnova
MAB2027
10707-456EA 428 USD
10707-456
Anti-CDKN2D Mouse Monoclonal Antibody [clone: DCS-100]
Antibodies
Mouse monoclonal antibody raised against full length recombinant CDKN2D.

Type: Primary
Antigen: CDKN2D
Clonality: Monoclonal
Clone: DCS-100
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR