Anti-POLI Rabbit Polyclonal Antibody

Supplier: Abnova
PAB12066
10685-798EA 345.2 USD
10685-798
Anti-POLI Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of POLI.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: POLI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR