Anti-LEFTY2 Rabbit Monoclonal Antibody [clone: 7C5G1H6H10]

Supplier: Abnova
MAB2032
10723-486EA 746.62 USD
10723-486
Anti-LEFTY2 Rabbit Monoclonal Antibody [clone: 7C5G1H6H10]
Antibodies
Rabbit monoclonal antibody raised against recombinant LEFTY2.

Recommended Dilutions: ELISA: 1:5000; Western Blot: 1:2000

Type: Primary
Antigen: LEFTY2
Clonality: Monoclonal
Clone: 7C5G1H6H10
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR