Anti-CD44 Mouse Monoclonal Antibody [clone: 5C10]

Supplier: Abnova
MAB2034
10706-570EA 329 USD
10706-570
Anti-CD44 Mouse Monoclonal Antibody [clone: 5C10]
Antibodies
Mouse monoclonal antibody raised against partial recombinant CD44.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: 5C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR