Anti-MAPK11 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB2785
10679-230EA 329 USD
10679-230
Anti-MAPK11 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of MAPK11.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:100-500; Immunohistochemistry: 1:50-100

Type: Primary
Antigen: MAPK11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR