Anti-TARDBP Mouse Monoclonal Antibody [clone: k1B8]

Supplier: Abnova
MAB2040
10740-524EA 329 USD
10740-524
Anti-TARDBP Mouse Monoclonal Antibody [clone: k1B8]
Antibodies
Mouse monoclonal antibody raised against partial recombinant TARDBP.

Type: Primary
Antigen: TARDBP
Clonality: Monoclonal
Clone: k1B8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR