Anti-UBE2L6 Mouse Monoclonal Antibody [clone: k1H3]

Supplier: Abnova
MAB2049
10743-622EA 329 USD
10743-622
Anti-UBE2L6 Mouse Monoclonal Antibody [clone: k1H3]
Antibodies
Mouse monoclonal antibody raised against partial recombinant UBE2L6.

Type: Primary
Antigen: UBE2L6
Clonality: Monoclonal
Clone: k1H3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR