Anti-D-Dimer Mouse Monoclonal Antibody [clone: DD41]

Supplier: Abnova
MAB2925
10676-138EA 345.2 USD
10676-138
Anti-D-Dimer Mouse Monoclonal Antibody [clone: DD41]
Antibodies
Mouse monoclonal antibody raised against D-Dimer.

Type: Primary
Antigen: D-Dimer
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: DD41
Epitope:
Host: IgG2a
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR