Anti-EPO Monoclonal Antibody [clone: Epo1]

Supplier: Abnova
MAB2937
10676-206EA 428.44 USD
10676-206
Anti-EPO Monoclonal Antibody [clone: Epo1]
Antibodies
Mouse monoclonal antibody raised against EPO.

Type: Primary
Antigen: EPO
Clonality: Monoclonal
Clone: Epo1
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR