Anti-FGA Mouse Monoclonal Antibody [clone: 15E11]

Supplier: Abnova
MAB2943
10676-272EA 530.32 USD
10676-272
Anti-FGA Mouse Monoclonal Antibody [clone: 15E11]
Antibodies
Mouse monoclonal antibody raised against Fibrinogen.

Type: Primary
Antigen: FGA
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 15E11
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR