Anti-FGA Mouse Monoclonal Antibody [clone: 15H12]

Supplier: Abnova
MAB2944
10676-274EA 530.32 USD
10676-274
Anti-FGA Mouse Monoclonal Antibody [clone: 15H12]
Antibodies
Mouse monoclonal antibody raised against Fibrinogen.

Type: Primary
Antigen: FGA
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 15H12
Epitope:
Host: IgG1
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR