Anti-FGA Mouse Monoclonal Antibody [clone: 1F7]

Supplier: Abnova
MAB2948
10676-256EA 428 USD
10676-256
Anti-FGA Mouse Monoclonal Antibody [clone: 1F7]
Antibodies
Mouse monoclonal antibody raised against synthetic peptide of FGA.

Type: Primary
Antigen: FGA
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 1F7
Epitope:
Host: IgG2a
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR