Anti-PRF1 Mouse Monoclonal Antibody [clone: 5B10]

Supplier: Abnova
MAB3715
10736-144EA 1170 USD
10736-144
Anti-PRF1 Mouse Monoclonal Antibody [clone: 5B10]
Antibodies
Mouse monoclonal antibody raised against partial recombinant PRF1.

Recommended Dilutions: Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 1:10

Type: Primary
Antigen: PRF1
Clonality: Monoclonal
Clone: 5B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR