Anti-DFFB Rabbit Polyclonal Antibody

Supplier: Abnova
PAB13074
10710-602EA 428 USD
10710-602
Anti-DFFB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of Dffb.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: DFFB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR