Anti-TNFSF15 Mouse Monoclonal Antibody [clone: L4G9]

Supplier: Abnova
MAB3731
10742-230EA 1061.25 USD
10742-230
Anti-TNFSF15 Mouse Monoclonal Antibody [clone: L4G9]
Antibodies
Mouse monoclonal antibody raised against recombinant TNFSF15.

Recommended Dilutions: Flow Cytometry: 1:1000

Type: Primary
Antigen: TNFSF15
Clonality: Monoclonal
Clone: L4G9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR