Anti-HSPA4 Mouse Monoclonal Antibody [clone: W27]

Supplier: Abnova
MAB3737
10718-176EA 1170 USD
10718-176
Anti-HSPA4 Mouse Monoclonal Antibody [clone: W27]
Antibodies
Mouse monoclonal antibody raised against native HSPA4.

Recommended Dilutions: Immunoprecipitation: 2 ?g/mg protein lysate; Western Blot: 0.5-1 ?g/ml; Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections): 2-4 ?g/ml

Type: Primary
Antigen: HSPA4
Clonality: Monoclonal
Clone: W27
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Human, Monkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR