Anti-C19ORF66 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB23888
10703-854EA 428.55 USD
10703-854
Anti-C19ORF66 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant C19orf66.

Recommended Dilutions: Immunohistochemistry: 1:500-1:1000; Western Blot: 1:250-1:500

Type: Primary
Antigen: C19ORF66
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR