Anti-HLA-DRA Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-12]

Supplier: Abnova
MAB4477
10717-556EA 329 USD
10717-556
Anti-HLA-DRA Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-12]
Antibodies
Mouse monoclonal antibody raised against HLA-DR.

Recommended Dilutions: Flow Cytometry: 10 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)

Type: Primary
Antigen: HLA-DRA
Clonality: Monoclonal
Clone: MEM-12
Conjugation: APC (Allophycocyanin)
Epitope:
Host:
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR