Anti-HLA-DPB1 Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-136]

Supplier: Abnova
MAB4485
10717-570EA 329 USD
10717-570
Anti-HLA-DPB1 Monoclonal Antibody (PE (Phycoerythrin)) [clone: MEM-136]
Antibodies
Mouse monoclonal antibody raised against HLA-DR/HLA-DP.

Recommended Dilutions: Flow Cytometry: 20 ?l in human blood cells 100 ?l in whole blood or 10^6 cells in a suspension)

Type: Primary
Antigen: HLA-DPB1
Clonality: Monoclonal
Clone: MEM-136
Conjugation: PE (Phycoerythrin)
Epitope:
Host:
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR