Anti-IgE Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BE5]

Supplier: Abnova
MAB4491
10718-758EA 329 USD
10718-758
Anti-IgE Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BE5]
Antibodies
Mouse monoclonal antibody raised against IgE.

Recommended Dilutions: Flow Cytometry: 5 ?g/ml

Type: Primary
Antigen: IgE
Clonality: Monoclonal
Clone: BE5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR