Anti-IgM Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: CH2]

Supplier: Abnova
MAB4495
10718-912EA 329 USD
10718-912
Anti-IgM Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: CH2]
Antibodies
Mouse monoclonal antibody raised against IgM.

Recommended Dilutions: Flow Cytometry: 2 ?g/ml

Type: Primary
Antigen: IgM
Clonality: Monoclonal
Clone: CH2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR