Anti-CD4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-241]

Supplier: Abnova
MAB4507
10706-508EA 329 USD
10706-508
Anti-CD4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEM-241]
Antibodies
Mouse monoclonal antibody raised against partial recombinant CD4.

Recommended Dilutions: Flow Cytometry: 10 ?g/ml

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: MEM-241
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR