Anti-STK31 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB2810
10682-356EA 406.15 USD
10682-356
Anti-STK31 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic peptide of STK31.

Recommended Dilutions: ELISA: 1:1000; Western Blot: 1:100-500; Immunohistochemistry: 1:50-100

Type: Primary
Antigen: STK31
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR