Anti-SMT3 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB11357
10735-650EA 746.62 USD
10735-650
Anti-SMT3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against recombinant SMT3.

Recommended Dilutions: ELISA: 1:1000-1:5000; Western Blot: 1:1000

Type: Primary
Antigen: SMT3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR